PRIH93ED86C8 Princeton BioMolecular Research, Inc.

(2Z)-2-(phenylhydrazono)propanoic acid

  • MDL number MFCD00984875
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart