PRIH93ED86D1 Princeton BioMolecular Research, Inc.

3,7,7-trimethylbicyclo[4.1.0]heptane

  • MDL number MFCD26099092
  • Molecular Weight : 138.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart