PRIH93ED86DA Princeton BioMolecular Research, Inc.

6-(4-isopropylphenyl)-2-phenyl-4,5-dihydropyridazin-3(2H)-one

  • MDL number MFCD02985313
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart