PRIH93ED86E3 Princeton BioMolecular Research, Inc.

N'-(3,4-dimethoxybenzyl)-N,N-dimethylsulfamide

  • MDL number MFCD01006045
  • Molecular Weight : 274.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart