PRIH93ED86EA Princeton BioMolecular Research, Inc.

methyl 2-[(4-nitrobenzyl)thio]benzoate

  • MDL number MFCD26103965
  • Molecular Weight : 303.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart