PRIH93ED86ED Princeton BioMolecular Research, Inc.

4-ethyl-5'-(4-nitrophenyl)-2,3'-diphenyl-2H,3'H-spiro[phthalazine-1,2'-[1,3,4]thiadiazole]

  • MDL number MFCD03614800
  • Molecular Weight : 505.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart