PRIH93ED86F2 Princeton BioMolecular Research, Inc.

1,1-dichlorocyclopropane

  • MDL number MFCD07783236
  • Molecular Weight : 110.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart