PRIH93ED8703 Princeton BioMolecular Research, Inc.

N-(2,5-dimethoxyphenyl)-N'-ethylthiourea

  • MDL number MFCD05408202
  • Molecular Weight : 240.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart