PRIH93ED8706 Princeton BioMolecular Research, Inc.

N-(2-cyclohex-1-en-1-ylethyl)-N'-phenylthiourea

  • MDL number MFCD01185575
  • Molecular Weight : 260.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart