PRIH93ED8716 Princeton BioMolecular Research, Inc.

N-1-naphthyl-2-nitrobenzamide

  • MDL number MFCD00578457
  • Molecular Weight : 292.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart