PRIH93ED8717 Princeton BioMolecular Research, Inc.

N-(4-fluorophenyl)-2-nitrobenzamide

  • MDL number MFCD00433008
  • Molecular Weight : 260.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart