PRIH93ED8724 Princeton BioMolecular Research, Inc.

diphenylmethyl imidothiocarbamate

  • MDL number MFCD31731655
  • Molecular Weight : 242.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart