PRIH93ED8736 Princeton BioMolecular Research, Inc.

2-[(1,3-benzodioxol-5-yloxy)acetyl]-N-methylhydrazinecarbothioamide

  • MDL number MFCD04243066
  • Molecular Weight : 283.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart