PRIH93ED8739 Princeton BioMolecular Research, Inc.

N-cyclohexyl-2-(phenylacetyl)hydrazinecarbothioamide

  • MDL number MFCD00708384
  • Molecular Weight : 291.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart