PRIH93ED873C Princeton BioMolecular Research, Inc.

N-allyl-2-(3-phenylpropanoyl)hydrazinecarbothioamide

  • MDL number MFCD03946353
  • Molecular Weight : 263.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart