PRIH93ED8759 Princeton BioMolecular Research, Inc.

(3E)-3-[(4-iodophenyl)imino]-1,3-dihydro-2H-indol-2-one

  • MDL number MFCD00442258
  • Molecular Weight : 348.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart