PRIH93ED8762 Princeton BioMolecular Research, Inc.

1-[(1Z)-2-nitroprop-1-en-1-yl]naphthalene

  • MDL number MFCD02090609
  • Molecular Weight : 213.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart