PRIH93ED8769 Princeton BioMolecular Research, Inc.

perylene-3,4,9,10-tetracarboxylic acid

  • MDL number MFCD00071725
  • Molecular Weight : 428.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart