PRIH93ED876B Princeton BioMolecular Research, Inc.

2-(3-ethyl-5-methylphenoxy)ethanamine

  • MDL number MFCD23712721
  • Molecular Weight : 179.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart