PRIH93ED8776 Princeton BioMolecular Research, Inc.

7-nitro-4,5-dihydro-1,4-benzoxazepin-3(2H)-one

  • MDL number MFCD03196005
  • Molecular Weight : 208.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart