PRIH93ED877D Princeton BioMolecular Research, Inc.

4-bromo-N-[3-(1,3-dioxan-2-yl)phenyl]benzamide

  • MDL number MFCD03196918
  • Molecular Weight : 362.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart