PRIH93ED8785 Princeton BioMolecular Research, Inc.

octahydro-1H-4,7-methanoinden-5-yl (3-chlorophenyl)carbamate

  • MDL number MFCD03196954
  • Molecular Weight : 305.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart