PRIH93ED8788 Princeton BioMolecular Research, Inc.

(2E)-2-cyano-N-(4-fluorophenyl)-3-(2-nitrophenyl)acrylamide

  • MDL number MFCD02298156
  • Molecular Weight : 311.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart