PRIH93ED878D Princeton BioMolecular Research, Inc.

N-cyclopropyl-2-nitro-5-[4-(4-phenyl-1,3-thiazol-2-yl)piperazin-1-yl]aniline

  • MDL number MFCD03197012
  • Molecular Weight : 421.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart