PRIH93ED8795 Princeton BioMolecular Research, Inc.

N,N-diethyl-5-methyl-4-oxo-2-[(2-oxo-2-{[3-(trifluoromethyl)phenyl]amino}ethyl)thio]-3-phenyl-3,4-dihydrothieno[2,3-d]pyrimidine-6-carboxamide

  • MDL number MFCD03197048
  • Molecular Weight : 574.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart