PRIH93ED8797 Princeton BioMolecular Research, Inc.

2-chloro-5-(methylthio)-N-(2-{[(2-phenoxyethyl)amino]carbonyl}phenyl)benzamide

  • MDL number MFCD03197061
  • Molecular Weight : 440.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart