PRIH93ED87A9 Princeton BioMolecular Research, Inc.

(2E)-3-(4,5-dimethoxy-2-nitrophenyl)-2-(2,2-dimethylpropanoyl)acrylonitrile

  • MDL number MFCD00170345
  • Molecular Weight : 318.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart