PRIH93ED87C7 Princeton BioMolecular Research, Inc.

1,3-diethyl-6-(morpholin-4-ylsulfonyl)quinazoline-2,4(1H,3H)-dione

  • MDL number MFCD03368316
  • Molecular Weight : 367.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart