PRIH93ED87CC Princeton BioMolecular Research, Inc.

4-tert-butyl-N-[3-(1,3-dioxan-2-yl)phenyl]benzenesulfonamide

  • MDL number MFCD03368353
  • Molecular Weight : 375.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart