PRIH93ED87D2 Princeton BioMolecular Research, Inc.

1-[(4-methylphenyl)amino]-3-(1,2,3,4-tetrahydro-9H-carbazol-9-yl)propan-2-ol

  • MDL number MFCD03368380
  • Molecular Weight : 334.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart