PRIH93ED87D9 Princeton BioMolecular Research, Inc.

methyl 4-({[5-(2-furyl)-4-phenyl-4H-1,2,4-triazol-3-yl]thio}methyl)benzoate

  • MDL number MFCD03153291
  • Molecular Weight : 391.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart