PRIH93ED87E5 Princeton BioMolecular Research, Inc.

2-{[7-(2H-1,2,3-benzotriazol-2-yl)-5-methyl-1,3-benzoxazol-2-yl]thio}-N-(tert-butyl)acetamide

  • MDL number MFCD03197348
  • Molecular Weight : 395.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart