PRIH93ED87E8 Princeton BioMolecular Research, Inc.

2-[(2-chloro-4,5-difluorobenzoyl)amino]-4,5-dimethylthiophene-3-carboxamide

  • MDL number MFCD03197383
  • Molecular Weight : 344.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart