PRIH93ED8809 Princeton BioMolecular Research, Inc.

2-(hexyloxy)-N-{5-[(4-phenylpiperidin-1-yl)methyl]-1,3-thiazol-2-yl}benzamide

  • MDL number MFCD03942227
  • Molecular Weight : 477.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart