PRIH93ED880C Princeton BioMolecular Research, Inc.

tetraphenyl piperazine-1,4-diylbis(phosphonate)

  • MDL number MFCD03765266
  • Molecular Weight : 550.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart