PRIH93ED8815 Princeton BioMolecular Research, Inc.

diphenyl (4-benzoylpiperazin-1-yl)phosphonate

  • MDL number MFCD03765433
  • Molecular Weight : 422.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart