PRIH93ED8824 Princeton BioMolecular Research, Inc.

4-methyl-N-[2-(piperidin-1-ylcarbonyl)phenyl]benzenesulfonamide

  • MDL number MFCD00376538
  • Molecular Weight : 358.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart