PRIH93ED8828 Princeton BioMolecular Research, Inc.

N-{2-[4-(pyrrolidin-1-ylsulfonyl)phenyl]ethyl}-1-naphthamide

  • MDL number MFCD02333898
  • Molecular Weight : 408.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart