PRIH93ED8848 Princeton BioMolecular Research, Inc.

2-chloro-4,5-difluoro-N-(4-{[(tetrahydrofuran-2-ylmethyl)amino]carbonyl}phenyl)benzamide

  • MDL number MFCD03765579
  • Molecular Weight : 394.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart