PRIH93ED884C Princeton BioMolecular Research, Inc.

N-cyclohexyl-7-nitrodibenzo[b,d]furan-2-sulfonamide

  • MDL number MFCD03942314
  • Molecular Weight : 374.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart