PRIH93ED884E Princeton BioMolecular Research, Inc.

N-(2,4-dimethoxyphenyl)-2-(4-methoxyphenyl)-4-methyl-1,2-dihydropyrimido[1,2-a]benzimidazole-3-carboxamide

  • MDL number MFCD03766479
  • Molecular Weight : 470.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart