PRIH93ED8862 Princeton BioMolecular Research, Inc.

(5Z,6E)-5,6-bis(phenylhydrazono)hexane-1,2,3,4-tetrol

  • MDL number MFCD00652466
  • Molecular Weight : 358.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart