PRIH93ED8866 Princeton BioMolecular Research, Inc.

N-[4-(4-chlorophenoxy)phenyl]-2-(1-naphthyl)acetamide

  • MDL number MFCD00175878
  • Molecular Weight : 387.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart