PRIH93ED886A Princeton BioMolecular Research, Inc.

2-[2-(2H-1,2,3-benzotriazol-2-yl)-4-methylphenoxy]-N-(2-phenoxyethyl)acetamide

  • MDL number MFCD02604183
  • Molecular Weight : 402.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart