PRIH93ED8873 Princeton BioMolecular Research, Inc.

4-(3,4-dimethoxyphenyl)-N-(4-ethoxyphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD00733377
  • Molecular Weight : 356.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart