PRIH93ED8876 Princeton BioMolecular Research, Inc.

2-(2-{4-[4-(4-bromophenyl)-1,3-thiazol-2-yl]piperazin-1-yl}ethyl)-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindole-1,3-dione

  • MDL number MFCD03819512
  • Molecular Weight : 513.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart