PRIH93ED888E Princeton BioMolecular Research, Inc.

4-methyl-N-(2-methyl-1,3-benzothiazol-6-yl)benzamide

  • MDL number MFCD01537945
  • Molecular Weight : 282.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart