PRIH93ED8890 Princeton BioMolecular Research, Inc.

3-[(5-anilino-4-phenyl-4H-1,2,4-triazol-3-yl)thio]butan-2-one

  • MDL number MFCD03819541
  • Molecular Weight : 338.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart