PRIH93ED8891 Princeton BioMolecular Research, Inc.

2-(4-methylphenyl)-3-phenyl-2,3-dihydropyrimido[4,5-b]quinolin-4(1H)-one

  • MDL number MFCD02915366
  • Molecular Weight : 365.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart