PRIH93ED88AC Princeton BioMolecular Research, Inc.

ethyl 6-tert-butyl-2-[(2-thienylcarbonyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

  • MDL number MFCD02920103
  • Molecular Weight : 391.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart